Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH3F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119793.5 | NP_195348.2 | 7 - 8 | 1352 | 122 | |
GenBank mRNA | BT030380.1 | - | 7 - 8 | 1063 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |