Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342408.1 | NP_001328464.1 | 4 - 5 | 1297 | 64 | |
NM_119801.4 | NP_568002.1 | 4 - 5 | 1359 | 64 | ||
GenBank mRNA | BT029220.1 | - | 4 - 5 | 1054 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |