Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119836.3 | NP_195390.2 | 5 - 6 | 394 | 72 | |
GenBank mRNA | BT012190.1 | - | 5 - 6 | 334 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |