Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HVA22K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119836.3 | NP_195390.2 | 7 - 8 | 569 | 73 | |
GenBank mRNA | BT012190.1 | - | 7 - 8 | 509 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |