Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036720.5 | NP_001031797.4 | 3 - 4 | 1898 | 90 | |
NM_001342426.1 | NP_001329338.1 | 2 - 3 | 1817 | 90 | ||
NM_001342427.1 | NP_001329337.1 | 2 - 3 | 1676 | 90 | ||
NM_119851.6 | NP_195405.4 | 2 - 3 | 1801 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |