Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MES9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119878.5 | NP_195432.1 | 1 - 2 | 578 | 78 | |
GenBank mRNA | AB493724.1 | - | 1 - 2 | 356 | 78 | |
AK117107.1 | - | 1 - 2 | 406 | 78 | ||
BT006227.1 | - | 1 - 2 | 356 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |