Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119927.3 | NP_195479.1 | 6 - 7 | 3104 | 92 | |
GenBank mRNA | AF025842.1 | - | 6 - 7 | 2887 | 92 | |
AY062484.1 | - | 6 - 7 | 2940 | 92 | ||
BT010592.1 | - | 6 - 7 | 2807 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |