Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119936.5 | NP_195488.2 | 1 - 2 | 720 | 80 | |
GenBank mRNA | AB493726.1 | - | 1 - 2 | 420 | 80 | |
AK176372.1 | - | 1 - 2 | 541 | 80 | ||
AK176432.1 | - | 1 - 2 | 470 | 80 | ||
AY102635.1 | - | 1 - 2 | 494 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |