Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179867.1 | NP_850198.1 | 2 - 3 | 1350 | 119 | |
NM_201856.4 | NP_973585.1 | 3 - 4 | 1071 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |