Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TKI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336615.1 | NP_001318368.1 | 6 - 7 | 823 | 76 | |
NM_179942.4 | NP_850273.1 | 7 - 8 | 1092 | 76 | ||
GenBank mRNA | AY064068.1 | - | 7 - 8 | 887 | 76 | |
AY096376.1 | - | 6 - 7 | 740 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |