Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129360.4 | NP_565880.1 | 7 - 8 | 1330 | 63 | |
NM_179960.1 | NP_850291.1 | 7 - 8 | 1244 | 63 | ||
GenBank mRNA | AF056969.1 | - | 7 - 8 | 1200 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |