Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YLS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125001.1 | NP_001118473.1 | 2 - 3 | 676 | 129 | |
NM_129443.2 | NP_030434.1 | 2 - 3 | 650 | 129 | ||
NM_179972.3 | NP_850303.1 | 2 - 3 | 809 | 129 | ||
GenBank mRNA | AB047808.1 | - | 2 - 3 | 656 | 129 | |
AY048272.1 | - | 2 - 3 | 673 | 129 | ||
AY084644.1 | - | 2 - 3 | 650 | 129 | ||
AY090381.1 | - | 2 - 3 | 649 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |