Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336762.1 | NP_001325386.1 | 5 - 6 | 1004 | 99 | |
NM_179982.3 | NP_850313.2 | 6 - 7 | 975 | 99 | ||
GenBank mRNA | AY576276.1 | - | 6 - 7 | 730 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |