Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100020.4 | NP_171645.1 | 2 - 3 | 373 | 95 | |
GenBank mRNA | AY519518.1 | - | 2 - 3 | 149 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |