Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331284.1 | NP_001322968.1 | 4 - 5 | 1142 | 66 | |
NM_100040.3 | NP_171664.1 | 4 - 5 | 929 | 66 | ||
GenBank mRNA | AY302057.1 | - | 4 - 5 | 742 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |