Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KEA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331307.1 | NP_001320631.1 | 18 - 19 | 3709 | 59 | |
NM_100062.5 | NP_171684.2 | 19 - 20 | 3733 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |