Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100065.5 | NP_563636.1 | 4 - 5 | 683 | 90 | |
GenBank mRNA | AB022861.1 | - | 4 - 5 | 405 | 90 | |
AF332441.1 | - | 3 - 4 | 341 | 90 | ||
AY064139.1 | - | 4 - 5 | 458 | 90 | ||
AY086844.1 | - | 4 - 5 | 458 | 90 | ||
AY097406.1 | - | 3 - 4 | 341 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |