Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331308.1 | NP_001322242.1 | 7 - 8 | 1107 | 124 | |
NM_100068.3 | NP_171690.1 | 8 - 9 | 997 | 124 | ||
GenBank mRNA | AF051326.1 | - | 8 - 9 | 962 | 124 | |
AK117412.1 | - | 8 - 9 | 969 | 124 | ||
BT005097.1 | - | 8 - 9 | 962 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |