Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160828.1 | NP_001154300.1 | 7 - 8 | 1196 | 78 | |
NM_001160829.1 | NP_001154301.1 | 7 - 8 | 1196 | 78 | ||
NM_001331317.1 | NP_001320318.1 | 6 - 7 | 1338 | 78 | ||
NM_100075.8 | NP_171697.3 | 7 - 8 | 1259 | 78 | ||
GenBank mRNA | AB290929.1 | - | 7 - 8 | 1108 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |