Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197959.1 | NP_001184888.1 | 3 - 4 | 290 | 75 | |
NM_001331318.1 | NP_001320381.1 | 3 - 4 | 459 | 75 | ||
NM_001331319.1 | NP_001320383.1 | 3 - 4 | 440 | 75 | ||
NM_001331320.1 | NP_001320382.1 | 2 - 3 | 524 | 75 | ||
NM_100081.5 | NP_563643.4 | 3 - 4 | 651 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |