Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RA-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100093.4 | NP_171715.1 | 2 - 3 | 220 | 111 | |
GenBank mRNA | AY052333.1 | - | 2 - 3 | 144 | 111 | |
AY061905.1 | - | 1 - 2 | 89 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |