Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RA-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100093.4 | NP_171715.1 | 4 - 5 | 316 | 95 | |
GenBank mRNA | AY052333.1 | - | 4 - 5 | 240 | 95 | |
AY061905.1 | - | 3 - 4 | 185 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |