Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331412.1 | NP_001321215.1 | 1 - 2 | 308 | 70 | |
NM_100227.6 | NP_171844.6 | 1 - 2 | 308 | 70 | ||
GenBank mRNA | AY372269.1 | - | 1 - 2 | 308 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |