Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POR C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035871.1 | NP_001030948.1 | 1 - 2 | 108 | 139 | |
NM_100243.4 | NP_171860.1 | 1 - 2 | 236 | 139 | ||
GenBank mRNA | AB035746.1 | - | 1 - 2 | 84 | 139 | |
AK226626.1 | - | 107 | 139 | |||
AY048263.1 | - | 1 - 2 | 102 | 139 | ||
AY088529.1 | - | 1 - 2 | 108 | 139 | ||
AY133569.1 | - | 1 - 2 | 60 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |