Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCI18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100270.4 | NP_563693.1 | 1 - 2 | 641 | 93 | |
GenBank mRNA | AK175162.1 | - | 1 - 2 | 526 | 93 | |
AK175560.1 | - | 1 - 2 | 608 | 93 | ||
AK176412.1 | - | 1 - 2 | 568 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |