Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331465.1 | NP_001321769.1 | 4 - 5 | 740 | 88 | |
NM_100297.3 | NP_171912.2 | 4 - 5 | 740 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |