Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF2 GAMMA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331466.1 | NP_001320247.1 | 7 - 8 | 1261 | 82 | |
NM_100298.3 | NP_171913.1 | 7 - 8 | 1095 | 82 | ||
GenBank mRNA | AF353097.1 | - | 7 - 8 | 940 | 82 | |
AY092973.1 | - | 7 - 8 | 1030 | 82 | ||
BT002095.1 | - | 6 - 7 | 832 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |