Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKL13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100324.3 | NP_171939.1 | 9 - 10 | 1369 | 107 | |
GenBank mRNA | AY943851.1 | - | 9 - 10 | 714 | 107 | |
BT015792.1 | - | 9 - 10 | 714 | 107 | ||
BT021997.1 | - | 9 - 10 | 803 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |