Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123762.2 | NP_001117234.1 | 4 - 5 | 660 | 89 | |
NM_100442.5 | NP_563745.1 | 5 - 6 | 922 | 89 | ||
GenBank mRNA | AY087913.1 | - | 5 - 6 | 636 | 89 | |
BT002049.1 | - | 5 - 6 | 614 | 89 | ||
BT008720.1 | - | 5 - 6 | 517 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |