Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CalS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331632.1 | NP_001322002.1 | 19 - 20 | 2704 | 74 | |
NM_100528.3 | NP_172136.2 | 19 - 20 | 2362 | 74 | ||
GenBank mRNA | HM049631.1 | - | 19 - 20 | 2320 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |