Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CalS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331632.1 | NP_001322002.1 | 26 - 27 | 3486 | 142 | |
NM_100528.3 | NP_172136.2 | 26 - 27 | 3144 | 142 | ||
GenBank mRNA | HM049631.1 | - | 26 - 27 | 3102 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |