Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100555.3 | NP_563771.1 | 3 - 4 | 518 | 112 | |
GenBank mRNA | AY045900.1 | - | 3 - 4 | 450 | 112 | |
AY091452.1 | - | 3 - 4 | 185 | 112 | ||
KJ139026.1 | - | 3 - 4 | 185 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |