Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112969.3 | NP_188714.2 | 17 - 18 | 1940 | 125 | |
GenBank mRNA | AJ297843.1 | - | 17 - 18 | 1762 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |