Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGLU17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125053.1 | NP_001118525.1 | 1 - 2 | 209 | 75 | |
NM_001337088.1 | NP_001324585.1 | 1 - 2 | 280 | 75 | ||
NM_001337089.1 | NP_001324587.1 | 1 - 2 | 280 | 75 | ||
NM_130011.3 | NP_181976.1 | 1 - 2 | 219 | 75 | ||
GenBank mRNA | AY074629.1 | - | 1 - 2 | 190 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |