Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337091.1 | NP_001323910.1 | 2 - 3 | 946 | 83 | |
NM_130021.3 | NP_181985.1 | 2 - 3 | 946 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |