Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337111.1 | NP_001318427.1 | 2 - 3 | 891 | 129 | |
NM_001337112.1 | NP_001324720.1 | 2 - 3 | 921 | 129 | ||
NM_130064.5 | NP_182026.5 | 2 - 3 | 944 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |