Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202819.1 | NP_001189748.1 | 2 - 3 | 781 | 84 | |
NM_130083.5 | NP_182045.1 | 2 - 3 | 634 | 84 | ||
GenBank mRNA | AF357529.1 | - | 2 - 3 | 383 | 84 | |
BT024623.1 | - | 2 - 3 | 328 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |