Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337170.1 | NP_001318433.1 | 6 - 7 | 910 | 160 | |
NM_001337171.1 | NP_001324870.1 | 5 - 6 | 1040 | 160 | ||
NM_001337172.1 | NP_001324871.1 | 6 - 7 | 783 | 160 | ||
NM_001337173.1 | NP_001324869.1 | 4 - 5 | 669 | 160 | ||
NM_130166.4 | NP_566062.1 | 5 - 6 | 539 | 160 | ||
GenBank mRNA | AK228245.1 | - | 5 - 6 | 494 | 160 | |
BT005208.1 | - | 5 - 6 | 416 | 160 | ||
DQ027021.1 | - | 5 - 6 | 416 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |