Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036471.1 | NP_001031548.1 | 3 - 4 | 527 | 82 | |
NM_130218.5 | NP_182176.2 | 4 - 5 | 825 | 82 | ||
NM_201972.1 | NP_973701.1 | 4 - 5 | 769 | 82 | ||
GenBank mRNA | AK229924.1 | - | 4 - 5 | 636 | 82 | |
AY669791.1 | - | 3 - 4 | 527 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |