Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337204.1 | NP_001324819.1 | 1 - 2 | 252 | 67 | |
NM_130222.2 | NP_182180.1 | 3 - 4 | 1107 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |