Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337212.1 | NP_001325093.1 | 4 - 5 | 864 | 112 | |
NM_001337213.1 | NP_001325092.1 | 3 - 4 | 951 | 112 | ||
NM_001337214.1 | NP_001325091.1 | 3 - 4 | 917 | 112 | ||
NM_130245.5 | NP_566085.1 | 4 - 5 | 864 | 112 | ||
NM_201974.3 | NP_973703.1 | 4 - 5 | 836 | 112 | ||
GenBank mRNA | AB046953.1 | - | 4 - 5 | 597 | 112 | |
AF272040.1 | - | 4 - 5 | 597 | 112 | ||
AK221652.1 | - | 4 - 5 | 709 | 112 | ||
AY099676.1 | - | 3 - 4 | 919 | 112 | ||
AY128856.1 | - | 1 - 2 | 126 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |