Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPa3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130253.3 | NP_182209.1 | 3 - 4 | 293 | 98 | |
GenBank mRNA | BT025278.1 | - | 2 - 3 | 233 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |