Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036478.2 | NP_001031555.1 | 7 - 8 | 710 | 64 | |
NM_001084605.1 | NP_001078074.1 | 7 - 8 | 710 | 64 | ||
NM_130315.5 | NP_182269.1 | 7 - 8 | 876 | 64 | ||
NM_201979.2 | NP_973708.1 | 7 - 8 | 710 | 64 | ||
GenBank mRNA | AF083688.1 | - | 7 - 8 | 664 | 64 | |
AJ271470.1 | - | 7 - 8 | 868 | 64 | ||
AK316899.2 | - | 7 - 8 | 696 | 64 | ||
AY074348.1 | - | 7 - 8 | 709 | 64 | ||
AY091388.1 | - | 7 - 8 | 627 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |