Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130326.3 | NP_182280.1 | 1 - 2 | 138 | 90 | |
GenBank mRNA | AY052663.1 | - | 1 - 2 | 134 | 90 | |
AY059875.1 | - | 1 - 2 | 138 | 90 | ||
AY093341.1 | - | 1 - 2 | 96 | 90 | ||
AY142070.1 | - | 1 - 2 | 96 | 90 | ||
Z49991.1 | - | 125 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |