Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130378.5 | NP_566125.4 | 12 - 13 | 2573 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |