Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100702.4 | NP_172306.1 | 1 - 2 | 787 | 115 | |
GenBank mRNA | AF254448.1 | - | 1 - 2 | 652 | 115 | |
AK118851.1 | - | 1 - 2 | 649 | 115 | ||
AY090918.1 | - | 1 - 2 | 590 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |