Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100725.4 | NP_563821.2 | 10 - 11 | 1827 | 72 | |
GenBank mRNA | AK226535.1 | - | 10 - 11 | 1712 | 72 | |
AX765812.1 | - | 10 - 11 | 1670 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |