Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACLA-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100814.4 | NP_172414.1 | 11 - 12 | 1343 | 76 | |
GenBank mRNA | AY127019.1 | - | 11 - 12 | 1235 | 76 | |
BT003017.1 | - | 11 - 12 | 1204 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |