Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100849.3 | NP_172448.1 | 3 - 4 | 2346 | 69 | |
GenBank mRNA | AY093057.1 | - | 3 - 4 | 2337 | 69 | |
BT008801.1 | - | 3 - 4 | 2176 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |