Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331921.1 | NP_001320481.1 | 8 - 9 | 2231 | 104 | |
NM_100944.2 | NP_172538.1 | 8 - 9 | 2374 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |